Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38880

Sigma-Aldrich

3-Methyl-2-butenoic acid

technical, ≥95% (T)

Synonym(s):

3,3-Dimethylacrylic acid, 3-Methylcrotonic acid, Senecioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCOOH
CAS Number:
Molecular Weight:
100.12
Beilstein/REAXYS Number:
1720305
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

assay

≥95% (T)

bp

194-195 °C (lit.)

mp

65-70 °C (lit.)
65-70 °C

SMILES string

C\C(C)=C\C(O)=O

InChI

1S/C5H8O2/c1-4(2)3-5(6)7/h3H,1-2H3,(H,6,7)

InChI key

YYPNJNDODFVZLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

200.1 °F - closed cup

flash_point_c

93.4 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service