Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38816

Sigma-Aldrich

4,4-Dimethoxytetrahydro-4H-pyran

purum, ≥97.0% (GC)

Synonym(s):

Tetrahydro-4H-pyran-4-one dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O3
CAS Number:
Molecular Weight:
146.18
Beilstein/REAXYS Number:
1236681
MDL number:
UNSPSC Code:
12352100

grade

purum

assay

≥97.0% (GC)

density

1.042 g/mL at 20 °C (lit.)

SMILES string

COC1(CCOCC1)OC

Other Notes

Reagent for protecting alcohols[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H.C.P. Roelen et al.
Rec. Trav. Chim., 106, 545-545 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service