38406
1,2:3,4-Di-O-isopropylidene-α-D-galactopyranose
purum, ≥97.0% (sum of enantiomers, GC)
Synonym(s):
1,2:3,4-Di-O-isopropylidene-D-galactopyranose
About This Item
grade
purum
assay
≥97.0% (sum of enantiomers, GC)
optical activity
[α]20/D −59±1°, c = 3% in chloroform
refractive index
n20/D 1.466 (lit.)
bp
117 °C/0.015 mmHg (lit.)
density
1.143 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CC1(C)O[C@H]2O[C@H](CO)[C@@H]3OC(C)(C)O[C@@H]3[C@H]2O1
InChI
1S/C12H20O6/c1-11(2)15-7-6(5-13)14-10-9(8(7)16-11)17-12(3,4)18-10/h6-10,13H,5H2,1-4H3/t6-,7+,8+,9-,10-/m1/s1
InChI key
POORJMIIHXHXAV-SOYHJAILSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service