Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38380

Sigma-Aldrich

Chlorodiisopropylsilane

purum, ≥98.0% (GC)

Synonym(s):

Diisopropylchlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2SiHCl
CAS Number:
Molecular Weight:
150.72
Beilstein/REAXYS Number:
2231355
MDL number:
UNSPSC Code:
12352000

grade

purum

assay

≥98.0% (GC)

refractive index

n20/D 1.429 (lit.)
n20/D 1.429

bp

58-60 °C/50 mmHg (lit.)

density

0.883 g/mL at 25 °C (lit.)

SMILES string

CC(C)[SiH](Cl)C(C)C

InChI

1S/C6H15ClSi/c1-5(2)8(7)6(3)4/h5-6,8H,1-4H3

InChI key

CGXYLRTYLIHXEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the stereoselective reduction of β-hydroxyketones[1]; Starting material for the preparation of diisopropyl-[5-dimethylamino(1-naphthyl)]silane, a fluorescent OH-selective protecting group reagent (and other diisopropylsilanes)[2]

replaced by

Product No.
Description
Pricing

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

100.4 °F - closed cup

flash_point_c

38 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L. Horner et al.
Journal of Organometallic Chemistry, 282, 175-175 (1985)
S. Anwar et al.
Journal of the Chemical Society. Chemical Communications, 831-831 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service