Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

383791

Sigma-Aldrich

N,N′-Bis(2,6-dimethylphenyl)perylene-3,4,9,10-tetracarboxylic diimide

Dye content 85 %

Synonym(s):

DXP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H26N2O4
CAS Number:
Molecular Weight:
598.65
Beilstein/REAXYS Number:
4626671
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 85%

mp

>300 °C (lit.)

λmax

526 nm

SMILES string

Cc1cccc(C)c1N2C(=O)c3ccc4c5ccc6C(=O)N(C(=O)c7ccc(c8ccc(C2=O)c3c48)c5c67)c9c(C)cccc9C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dyes and Pigments, 11, 303-303 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service