Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38305

Sigma-Aldrich

Diisopropylamine dihydrofluoride

purum, ≥98.0% (F)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NH · 2HF
CAS Number:
Molecular Weight:
141.20
Beilstein/REAXYS Number:
8913832
MDL number:
UNSPSC Code:
12352101

grade

purum

assay

≥98.0% (F)

SMILES string

F.[F-].CC(C)[NH2+]C(C)C

Application

Fluorinating agent

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service