Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36870

Sigma-Aldrich

2,4-Difluoroaniline

purum, ≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NH2
CAS Number:
Molecular Weight:
129.11
Beilstein/REAXYS Number:
2802556
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥95.0% (GC)

refractive index

n20/D 1.506 (lit.)
n20/D 1.506

bp

170 °C/753 mmHg (lit.)

mp

−7.5 °C (lit.)

density

1.268 g/mL at 25 °C (lit.)

SMILES string

Nc1ccc(F)cc1F

InChI

1S/C6H5F2N/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

CEPCPXLLFXPZGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Chih Huang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 176-179 (2012-04-07)
We applied the two-color resonant two-photon ionization and mass-analyzed threshold ionization techniques to record the vibronic and cation spectra of 2,4-difluoroaniline. The cation spectra were recorded by ionizing via the 0(0), X(1), 6b(1), and 1(1) levels of the electronically excited
Sergey E Korchak et al.
The Journal of chemical physics, 137(9), 094503-094503 (2012-09-11)
Effects of spin-spin interactions on the nuclear magnetic relaxation dispersion (NMRD) of protons were studied in a situation where spin ½ hetero-nuclei are present in the molecule. As in earlier works [K. L. Ivanov, A. V. Yurkovskaya, and H.-M. Vieth
Duoli Guo et al.
AAPS PharmSciTech, 13(2), 661-673 (2012-05-04)
A stability-indicating high-performance liquid chromatography method to quantify 2-(2,4-difluorophenyl)-4,5,6,7-tetrafluoroisoindoline-1,3-dione (NSC-726796) and its three main degradation products was developed. This method was used to investigate its degradation kinetics and mechanism. The reaction follows first-order kinetics and appears to be base catalyzed
Gergely Rácz et al.
Pathology oncology research : POR, 18(3), 579-584 (2011-12-14)
Disinfection of raw water is essential to the production of drinking water. However, by-products of disinfection may exert toxic effects. The potential toxic effects of two of these compounds, 4-ethylbenzaldehyde (EBA) and 2,4-difluoroaniline (DFA) were investigated using the zebrafish (Danio
P J Boogaard et al.
Environmental health perspectives, 102 Suppl 6, 27-29 (1994-10-01)
Exposure to 2,4-difluoroaniline (DFA) was monitored by GC-MS of DFA adducts bound to hemoglobin (Hb). In two studies, involving 20 and 16 workers potentially exposed to low concentrations of DFA, median concentrations of 10 (range 1-83) and 20 (range 4-322)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service