Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36708

Supelco

1,3-Dichlorobenzene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein/REAXYS Number:
956618
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

vapor pressure

5 mmHg ( 38.8 °C)

product line

PESTANAL®

assay

98% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.546 (lit.)

bp

172-173 °C (lit.)

mp

−25-−22 °C (lit.)

density

1.288 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1cccc(Cl)c1

InChI

1S/C6H4Cl2/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

ZPQOPVIELGIULI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

152.6 °F - closed cup

flash_point_c

67.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Yoshida et al.
Analytical chemistry, 73(18), 4417-4421 (2001-09-29)
A distributed-feedback dye laser has been developed for achieving the efficient multiphoton ionization of chlorobenzene and dichlorobenzene, that is, precursor molecules of dioxins. This tunable picosecond laser with a narrow spectral line width, that is, a nearly transform-limited pulse, provides
J K Bibler-Muckelbauer et al.
Virology, 202(1), 360-369 (1994-07-01)
Crystallographic studies of human rhinovirus 14 (HRV14) crystals soaked with fragments of antiviral WIN compounds, at high concentrations (82-200 micrograms/ml), show the compounds bind into the hydrophobic beta-barrel (WIN pocket) of VP1. Two of these short compounds (5-[3,5-dimethyl-4-hydroxyphenyl]-2-methyltetrazole and phenol
Michael Bunge et al.
Environmental microbiology, 10(10), 2670-2683 (2008-07-17)
Enrichment cultures capable of reductively dechlorinating 1,2,4-trichlorodibenzo-p-dioxin (1,2,4-TrCDD) were shown to dechlorinate 1,2,3-trichlorobenzene (1,2,3-TrCB) to 1,3-dichlorobenzene. To test if this activity can be used to enrich for dioxin-dechlorinating bacteria, a two-liquid phase cultivation with 200 mM 1,2,3-TrCB dissolved in hexadecane
T Umemura et al.
Toxicology and applied pharmacology, 137(2), 268-274 (1996-04-01)
The acute hepatotoxicity of isomers of dichlorobenzene (o-, m-, and p-DCB) was compared in livers of male B6C3F1 mice at different time points after a single intragastric administration. The highest doses of o-, m-, and p-DCB administered, 300, 300, and
D Mahadevan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 962-969 (2011-05-13)
The FT-IR and FT-Raman vibrational spectra of 1,3-dichlorobenzene (1,3-DCB) have been recorded using Bruker IFS 66 V Spectrometer in the range 4000-100 cm(-1). A detailed vibrational spectral analysis has been carried out and assignments of the observed fundamental bands have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service