Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

36169

Supelco

MGK 326

PESTANAL®, analytical standard

Synonym(s):

Dipropyl pyridine-2,5-dicarboxylate, Dipropylisocinchomeronate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO4
CAS Number:
Molecular Weight:
251.28
Beilstein/REAXYS Number:
229600
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCOC(=O)c1ccc(nc1)C(=O)OCCC

InChI

1S/C13H17NO4/c1-3-7-17-12(15)10-5-6-11(14-9-10)13(16)18-8-4-2/h5-6,9H,3-4,7-8H2,1-2H3

InChI key

IITCWRFYJWUUPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

MGK 326 may be used as an analytical reference standard for the determination of the analyte in pyrethrum extracts by reversed-phase high-performance liquid chromatography with diode-array detection.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

200.1 °F - closed cup

flash_point_c

93.4 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas J Begg et al.
Veterinary microbiology, 219, 53-62 (2018-05-21)
Johne's disease (JD) or paratuberculosis is an economically significant, chronic enteropathy of ruminants caused by Mycobacterium avium subspecies paratuberculosis (MAP). Experimental models of JD in cattle are logistically challenging due to the need for long term monitoring, because the clinical
Lin Zhao et al.
Molecular medicine reports, 22(1), 436-444 (2020-05-08)
Posterior capsular opacification (PCO) is the main complication following cataract surgery. The proliferation of the residual lens epithelial cells (LECs) serves an important role in PCO formation. The authors' previous study revealed that vitamin C inhibited the proliferation of human
Direct determination of pyrethrins in pyrethrum extracts by reversed-phase high-performance liquid chromatography with diode-array detection.
Wang IH, et al.
Journal of Chromatography A, 766(1-2), 277-281 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service