Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34557

Supelco

Paraquat diiodide-(dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):

1,1′-Di(methyl-d3)-4,4′-bipyridinium diiodide, Methyl viologen-d6 diiodide, Paraquat-d6 diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D6H8I2N2
Molecular Weight:
446.10
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])[N+](C=C1)=CC=C1C2=CC=[N+](C([2H])([2H])[2H])C=C2

General description

Paraquat diiodide-(dimethyl-d6) is a deuterated analog of bipyridylium herbicide paraquat, wherein the dimethyl protons are replaced by deuterium.[1]

Application

Paraquat diiodide-(dimethyl-d6) may be used as a deuterated internal standard to quantify paraquat in cowpeas by ultra-performance liquid chromatography coupled to tandem mass spectrometry (UPLC-MS/MS).[1]
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of paraquat and diquat: LC-MS method optimization and validation
Pizzutti IR, et al.
Food Chemistry, 209, 248-255 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service