34527
2,3,5-Trimethacarb solution
10 μg/mL in cyclohexane, PESTANAL®, analytical standard
About This Item
grade
analytical standard
product line
PESTANAL®
concentration
10 μg/mL in cyclohexane
storage temp.
2-8°C
SMILES string
CC1=CC(C)=C(C)C(OC(NC)=O)=C1
InChI
1S/C11H15NO2/c1-7-5-8(2)9(3)10(6-7)14-11(13)12-4/h5-6H,1-4H3,(H,12,13)
InChI key
NYOKZHDTNBDPOB-UHFFFAOYSA-N
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
-0.4 °F
flash_point_c
-18 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service