Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33804

Sigma-Aldrich

1,3-Dibenzyloxy-2-propanol

purum, ≥97.0% (GC)

Synonym(s):

1,3-Bis(benzyloxy)-2-propanol, 1,3-Di-O-benzylglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2OCH2)2CHOH
CAS Number:
Molecular Weight:
272.34
Beilstein/REAXYS Number:
1986774
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (GC)

refractive index

n20/D 1.549 (lit.)
n20/D 1.549

bp

226-227 °C/3 mmHg (lit.)

density

1.103 g/mL at 20 °C (lit.)

SMILES string

OC(COCc1ccccc1)COCc2ccccc2

InChI

1S/C17H20O3/c18-17(13-19-11-15-7-3-1-4-8-15)14-20-12-16-9-5-2-6-10-16/h1-10,17-18H,11-14H2

InChI key

ARLSYSVVBAMYKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Dibenzyloxy-2-propanol was used as starting reagent in the synthesis of difluorinated cyclopropane nucleoside analogs possessing both two vicinal hydroxymethyl groups and a methylene spacer between the base and the ring. It was used as starting reagent in the synthesis of ganciclovir (GCV, 9-[(1,3-dihydroxy-2-propoxy)methyl]-guanine). It was also used in the synthesis of [3H] endocannabinoid 2-arachidonoylglycerol.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M MacCarrone et al.
European journal of biochemistry, 268(3), 819-825 (2001-02-13)
The endocannabinoid 2-arachidonoylglycerol (2-Delta(4)Ach-Gro) activates human platelets in platelet-rich plasma at physiological concentrations. The activation was inhibited by selective antagonists of CB(1) and CB(2) cannabinoid receptors, but not by acetylsalicylic acid. Human platelets can metabolize 2-Delta(4)Ach-Gro by internalization through a
Preparation of novel difluorocyclopropane nucleosides.
Csuk R and Thiede G.
Tetrahedron, 55(3), 739-750 (1999)
Ji-Quan Wang et al.
Bioorganic & medicinal chemistry letters, 13(22), 3933-3938 (2003-11-01)
Positron emission tomography (PET) herpes simplex virus thymidine kinase (HSV-tk) gene reporter probes 9-[(3-[(18)F]fluoro-1-hydroxy-2-propoxy)methyl]guanine ([(18)F]FHPG) and 9-(4-[(18)F]fluoro-3-hydroxymethylbutyl)guanine ([(18)F]FHBG) were prepared by nucleophilic substitution of the appropriate tosylated precursors with [(18)F]KF/Kryptofix 2.2.2 followed by a quick deprotection reaction and purification with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service