32729
Febantel-d6
VETRANAL®, analytical standard
Synonym(s):
N-{2-[2,3-Bis-(methoxy-d3-carbonyl)-guanido]-5-(phenylthio)-phenyl}-2-methoxyacetamide
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
clinical
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
mass shift
M+6
SMILES string
[2H]C([2H])([2H])OC(=O)N\C(NC(=O)OC([2H])([2H])[2H])=N/c1ccc(Sc2ccccc2)cc1NC(=O)COC
InChI
1S/C20H22N4O6S/c1-28-12-17(25)21-16-11-14(31-13-7-5-4-6-8-13)9-10-15(16)22-18(23-19(26)29-2)24-20(27)30-3/h4-11H,12H2,1-3H3,(H,21,25)(H2,22,23,24,26,27)/i2D3,3D3
InChI key
HMCCXLBXIJMERM-XERRXZQWSA-N
General description
application
Legal Information
related product
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service