Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32621

Sigma-Aldrich

Diallyl disulfide

technical, ~80% (GC)

Synonym(s):

Allyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2SSCH2CH=CH2
CAS Number:
Molecular Weight:
146.27
Beilstein/REAXYS Number:
1699241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

assay

~80% (GC)

impurities

10-20% diallyl sulfide

refractive index

n20/D 1.54

density

1.0 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCSSCC=C

InChI

1S/C6H10S2/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-6H2

InChI key

PFRGXCVKLLPLIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Yi Chen et al.
Archives of toxicology, 86(2), 231-238 (2011-09-01)
Diallyl disulfide (DADS), one of the major organosulfur compounds of garlic, is recognized as a group of potential chemopreventive compounds. In this study, we examines the early signaling effects of DADS on human colorectal cancer cells SW480 loaded with Ca(2+)-sensitive
Mohammed O Altonsy et al.
Nutrition and cancer, 64(8), 1251-1260 (2012-11-21)
The health benefits of garlic have been proven by epidemiological and experimental studies. Diallyl disulphide (DADS), the major organosulfur compound found in garlic oil, is known to lower the incidence of breast cancer both in vitro and in vivo. The
Akeem O Lawal et al.
Environmental toxicology and pharmacology, 32(2), 266-274 (2011-08-17)
Garlic has been reported in many previous studies as a potent chemopreventive agent. The protective effect of garlic has been ascribed to the presence of organosulphur compounds (OSC). In this study, the efficacy of aged garlic extract (AGE) compared to
Hyun Soo Park et al.
Journal of food science, 76(4), T105-T111 (2012-03-16)
The effect of diallyl disulfide (DADS), a major component of an oil-soluble allyl sulfide garlic (Allium sativum) derivative, on the correlation between anti-invasive activity and tightening of tight junctions (TJs) was investigated in human gastric adenocarcinoma AGS cells. Our data
Laura Masuelli et al.
Frontiers in bioscience (Landmark edition), 17, 498-508 (2011-12-29)
Malignant tumors of mesenchimal origin such as rhabdomyosarcoma and osteosarcoma are highly aggressive pedriatic malignancies with a poor prognosis. Indeed, the initial response to chemotherapy is followed by chemoresistance. Diallyl disulfide (DADS), resveratrol (RES) and curcumin (CUR) are dietary chemopreventive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service