32531
Tralomethrin
PESTANAL®, analytical standard, mixture of diastereomers
Synonym(s):
(S)-α-Cyano-3-phenoxybenzyl (1R,3S)-2,2-dimethyl-3-[(RS)-1,2,2,2-tetrabromoethyl]cyclopropanecarboxylate
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
storage temp.
2-8°C
SMILES string
O=C(OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1)C3C(C(Br)C(Br)(Br)Br)C3(C)C
InChI
1S/C22H19Br4NO3/c1-21(2)17(19(23)22(24,25)26)18(21)20(28)30-16(12-27)13-7-6-10-15(11-13)29-14-8-4-3-5-9-14/h3-11,16-19H,1-2H3
InChI key
YWSCPYYRJXKUDB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Tralomethrin is not authorized for use in the European Union as per the European Council Directive 91/414/EEC concerning the placement of plant protection products on the European market. A default maximum residue level (MRL) of 0.01 mg/kg has been set according to Reg 396/2005
Application
- Evaluate the analytical behavior of tralomethrin using GC-ECD and GC-MS, and compare the analytical parameters for tralomethrin and deltamethrin
- Evaluate the residue levels of pyrifenox, pyridaben, and tralomethrin in tomatoes grown in a plastic greenhouse, and assess the effect of three different household processes and the "unit to unit" variability of these pesticides in tomatoes
- Study the electrochemical reduction behavior of tralomethrin and tralocythrin at glassy carbon and mercury electrodes in the non-aqueous solvents acetonitrile, and methanol
- Evaluate the degradation behavior and residue levels of pyridaben and tralomethrin in peppers grown in a plastic greenhouse
- Study the association between environmental exposure to pesticides and risk of epilepsy
- Determine the fate and persistence of tralomethrin and deltamethrin in simulated outdoor pond microcosms
Legal Information
Not finding the right product?
Try our Product Selector Tool.
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service