32433
Primisulfuron-methyl
PESTANAL®, analytical standard
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
COC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(OC(F)F)cc(OC(F)F)n2
InChI
1S/C15H12F4N4O7S/c1-28-11(24)7-4-2-3-5-8(7)31(26,27)23-15(25)22-14-20-9(29-12(16)17)6-10(21-14)30-13(18)19/h2-6,12-13H,1H3,(H2,20,21,22,23,25)
Inchi Key
ZTYVMAQSHCZXLF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Surface and groundwater samples by solid-phase extraction (SPE) and high performance liquid chromatography-electrospray ionization-mass spectrometry (HPLC/ESI-MS).
- Tap and leaching water samples by SPE and LC coupled to tandem mass spectrometry (MS/MS) with ESI and selected reaction monitoring (SRM) detection.
- Wine samples by vortex-assisted ionic liquid dispersive liquid–liquid microextraction ((VA-IL-DLLME)) and capillary HPLC.
Legal Information
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service