Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32348

Supelco

Cyhalofop

PESTANAL®, analytical standard

Synonym(s):

(2R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propanoic acid, (R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propionic acid, Cyhalofop acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12FNO4
CAS Number:
Molecular Weight:
301.27
Beilstein/REAXYS Number:
11324819
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C[C@H](C(O)=O)OC1=CC=C(OC2=C(F)C=C(C#N)C=C2)C=C1

InChI

1S/C16H12FNO4/c1-10(16(19)20)21-12-3-5-13(6-4-12)22-15-7-2-11(9-18)8-14(15)17/h2-8,10H,1H3,(H,19,20)/t10-/m1/s1

InChI key

ROBSGBGTWRRYSK-SNVBAGLBSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

pcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service