Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

310271

Sigma-Aldrich

Phosphoric acid

reagent grade, crystalline, ≥98%

Synonym(s):

Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein/REAXYS Number:
1921286
EC Number:
MDL number:
UNSPSC Code:
12352106

grade

reagent grade

vapor density

3.4 (vs air)

vapor pressure

0.03 mmHg ( 20 °C)
2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)
5 mmHg ( 25 °C)

assay

≥98%

form

crystalline

bp

158 °C (lit.)
158 °C/760 mmHg

mp

~40 °C (lit.)
41-44 °C

density

1.685 g/mL at 25 °C (lit.)

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

Inchi Key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessio Ciulli et al.
Journal of medicinal chemistry, 49(16), 4992-5000 (2006-08-04)
Mapping interactions at protein-ligand binding sites is an important aspect of understanding many biological reactions and a key part of drug design. In this paper, we have used a fragment-based approach to probe "hot spots" at the cofactor-binding site of
Alex W Gregory et al.
Organic letters, 15(17), 4330-4333 (2013-08-30)
A highly enantioselective hydroamination/N-sulfonyliminium cyclization cascade is reported using a combination of gold(I) and chiral phosphoric acid catalysts. An initial 5-exo-dig hydroamination and a subsequent phosphoric acid catalyzed cyclization process provide access to complex sulfonamide scaffolds in excellent yield and
W M Fennis et al.
Journal of dental research, 93(1), 36-41 (2013-10-25)
The objective of this randomized control trial was to compare the five-year clinical performance of direct and indirect resin composite restorations replacing cusps. In 157 patients, 176 restorations were made to restore maxillary premolars with Class II cavities and one
Shuliang Liu et al.
Journal of immunology (Baltimore, Md. : 1950), 183(9), 5537-5547 (2009-10-09)
Citrobacter koseri is a Gram-negative bacterium that can cause a highly aggressive form of neonatal meningitis, which often progresses to establish multifocal brain abscesses. Despite its tropism for the brain parenchyma, microglial responses to C. koseri have not yet been
Li Yan Chan et al.
The Journal of organic chemistry, 78(17), 8826-8832 (2013-08-07)
A simple and efficient method is developed for Pd-catalyzed ortho-acetoxylation using organophosphates, namely, benzylic phosphonic and aryl phosphoric monoacids, as the directing group.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service