Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29200

Sigma-Aldrich

cis,cis-1,3,5-Cyclohexanetriol dihydrate

purum, ≥97.5% (GC)

Synonym(s):

cis,cis-1,3,5-Trihydroxycyclohexane, cis-Phloroglucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O3 · 2H2O
CAS Number:
Molecular Weight:
168.19
Beilstein/REAXYS Number:
2203732
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

purum

assay

≥97.5% (GC)

mp

108-111 °C (lit.)
108-111 °C

SMILES string

O.O.O[C@H]1C[C@@H](O)C[C@@H](O)C1

InChI

1S/C6H12O3.2H2O/c7-4-1-5(8)3-6(9)2-4;;/h4-9H,1-3H2;2*1H2/t4-,5+,6-;;

InChI key

MAPLMYMZLRIJSP-XHTDGWPASA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Triol used for the protection of carboxylic acids as orthoesters which are stable to Grignard reagents[1][2]; Building block used in stereoselective syntheses[3][4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F. Prasad, O. Repic
Tetrahedron Letters, 25, 4889-4889 (1984)
F. Bohlmann et al.
Chemische Berichte, 97, 1839-1839 (1964)
A novel synthesis of prostaglandin F.
R B Woodward et al.
Journal of the American Chemical Society, 95(20), 6853-6855 (1973-10-03)
G. Voss et al.
Helvetica Chimica Acta, 66, 2294-2294 (1983)
Lisa Gasperini et al.
Antioxidants & redox signaling, 22(9), 772-784 (2014-12-10)
Several neurodegenerative disorders show alterations in glutamatergic synapses and increased susceptibility to excitotoxicity. Mounting evidence suggests a central role for the cellular prion protein (PrP(C)) in neuroprotection. Therefore, the loss of PrP(C) function occurring in prion disorders may contribute to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service