Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28225

Sigma-Aldrich

Disperse Yellow 3

for microscopy

Synonym(s):

4-(2-Hydroxy-5-methylphenylazo)acetanilide, N-[4-(2-Hydroxy-5-methylphenylazo)phenyl]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15N3O2
CAS Number:
Molecular Weight:
269.30
Colour Index Number:
11855
Beilstein/REAXYS Number:
1915225
EC Number:
MDL number:
UNSPSC Code:
12171500

grade

for microscopy

mp

268-270 °C (lit.)

application(s)

environmental

SMILES string

CC(=O)Nc1ccc(cc1)N=Nc2cc(C)ccc2O

InChI

1S/C15H15N3O2/c1-10-3-8-15(20)14(9-10)18-17-13-6-4-12(5-7-13)16-11(2)19/h3-9,20H,1-2H3,(H,16,19)

InChI key

PXOZAFXVEWKXED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Dye standard for the assay of allergy-inducing dyes in textiles[1][2]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francine Inforçato Vacchi et al.
The Science of the total environment, 601-602, 230-236 (2017-05-30)
The combination of chemical analyses and bioassays allows the identification of potentially mutagenic compounds in different types of samples. Dyes can be considered as emergent contaminants and were detected in waters, under the influence of textile activities. The objective of
K.L. Hatch
Textile Res., 54, 664-664 (1984)
K L Hatch et al.
Journal of the American Academy of Dermatology, 32(4), 631-639 (1995-04-01)
The literature concerning textile dye dermatitis published during the last decade was reviewed. Sixty-one cases of dye-allergic contact dermatitis in which the presentation or course of the dermatitis was unusual or the dye allergen was one not previously reported have
Davide Tamburro et al.
Journal of the American Chemical Society, 133(47), 19178-19188 (2011-10-18)
Many low-abundance biomarkers for early detection of cancer and other diseases are invisible to mass spectrometry because they exist in body fluids in very low concentrations, are masked by high-abundance proteins such as albumin and immunoglobulins, and are very labile.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service