Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27028

Sigma-Aldrich

Sodium cholate hydrate

≥96.0% (dry matter, NT)

Synonym(s):

3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt, Cholalic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NaO5 · xH2O
CAS Number:
Molecular Weight:
430.55 (anhydrous basis)
Beilstein/REAXYS Number:
3582354
EC Number:
MDL number:
UNSPSC Code:
12352200

description

anionic

assay

≥96.0% (dry matter, NT)

optical activity

[α]20/D +35±2°, c = 0.6% in ethanol (dry matter)

mol wt

micellar avg mol wt 900-1300

aggregation number

2-3

impurities

~4% water

CMC

9-15 mM (20-25°C)

solubility

H2O: 0.1 M, clear

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

HLB

18

SMILES string

O.[Na+].C[C@H](CCC([O-])=O)C1CCC2C3[C@H](O)CC4C[C@H](O)CC[C@]4(C)C3C[C@H](O)[C@]12C

InChI

1S/C24H40O5.Na.H2O/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26;;/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29);;1H2/q;+1;/p-1/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-;;/m1../s1

InChI key

MUVVIYFKOVLQHL-RCVKHMDESA-M

Looking for similar products? Visit Product Comparison Guide

application

Sodium cholate hydrate was used in the sonication and purification of single-walled carbon nanotubes prior to electrophoresis.[1][2]

Biochem/physiol Actions

Sodium cholate is an anionic detergent. It forms aggregates or micelles in aqueous solution.[3] It slightly increases the solubility of insoluble molecules such as cholesterol, phospholipids and fatty acids.[4]

Preparation Note

Sodium cholate hydrate yields clear, colorless solution in water (0.1 M).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Shefer et al.
Hepatology (Baltimore, Md.), 22(4 Pt 1), 1215-1221 (1995-10-01)
We examined the effects of feeding deoxycholic acid (1% and 0.4% of diet), alone and in combination with ursodeoxycholic acid, on serum and biliary bile acid concentrations, hepatic morphology, and the activities and steady-state messenger RNA (mRNA) levels of HMG-CoA
D K Spady et al.
The Journal of biological chemistry, 267(8), 5584-5591 (1992-03-15)
In vivo regulation of hepatic sterol metabolism was examined in the rat. Sodium cholate markedly suppressed hepatic 7 alpha-hydroxylase mRNA levels and activity when fed to rats on a low cholesterol diet. Sterol balance was maintained solely by decreasing hepatic
Giulio Lolli et al.
The journal of physical chemistry. B, 110(5), 2108-2115 (2006-02-14)
The (n,m) population distribution of single-walled carbon nanotubes obtained on supported CoMo catalysts has been determined by photoluminescence and optical absorption. It has been found that the (n,m) distribution can be controlled by varying the gaseous feed composition, the reaction
Daniel A Heller et al.
Journal of the American Chemical Society, 126(44), 14567-14573 (2004-11-04)
Gel electrophoresis and column chromatography conducted on individually dispersed, ultrasonicated single-walled carbon nanotubes yield simultaneous separation by tube length and diameter. Electroelution after electrophoresis is shown to produce highly resolved fractions of nanotubes with average lengths between 92 and 435
Naomichi Nishimura et al.
Journal of nutritional science and vitaminology, 49(1), 21-26 (2003-07-29)
The effects of taurine on serum cholesterol levels and hepatic cholesterol 7alpha-hydroxylase activity (CYP7A1) were studied in rats fed cholestyramine or high amounts of sodium cholate in order to alter the intestinal pool of bile acids. Rats were fed a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service