Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26667

Sigma-Aldrich

3-Chlorocinnamic acid

purum, ≥99.0% (T)

Synonym(s):

trans-3-(3-Chlorophenyl)propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
182.60
Beilstein/REAXYS Number:
2207378
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

assay

≥99.0% (T)

mp

161-164 °C (lit.)
161-164 °C

SMILES string

OC(=O)\C=C\c1cccc(Cl)c1

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service