Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26035

Sigma-Aldrich

9-Chloro-9-phenylxanthene

purum, ≥97.0% (AT)

Synonym(s):

Pixyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13ClO
CAS Number:
Molecular Weight:
292.76
Beilstein/REAXYS Number:
237712
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (AT)

mp

102-106 °C (lit.)
102-106 °C

SMILES string

ClC2(c1ccccc1)c3ccccc3Oc4ccccc24

InChI

1S/C19H13ClO/c20-19(14-8-2-1-3-9-14)15-10-4-6-12-17(15)21-18-13-7-5-11-16(18)19/h1-13H

InChI key

HTGMODSTGYKJDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the introduction of the 9-phenyl-9-xanthenyl protecting group for alcohols[1][2][3]; N-Protection of carboxamides[4]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Gioeli et al.
Tetrahedron Letters, 22, 969-969 (1981)
H. Echner and W. Voelter et al.
Proc. 22nd Eur. Pept. Symp.: Peptides 1992 null
J.B. Chattopadhyaya et al.
Journal of the Chemical Society. Chemical Communications, 639-639 (1978)
N. Balgobin et al.
Tetrahedron Letters, 22, 1915-1915 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service