Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24791

Sigma-Aldrich

6-Chloro-1-hexanol

purum, ≥95.0% (GC)

Synonym(s):

1-Chloro-6-hydroxyhexane, Hexamethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)6OH
CAS Number:
Molecular Weight:
136.62
Beilstein/REAXYS Number:
1697307
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

assay

≥95.0% (GC)

refractive index

n20/D 1.455

bp

108-112 °C/14 mmHg (lit.)

density

1.024 g/mL at 20 °C (lit.)

SMILES string

OCCCCCCCl

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service