Skip to Content
MilliporeSigma
All Photos(1)

Documents

22477

Supelco

Chelex® 20 sodium form

ion exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

loss

~80% loss on drying, 110 °C

matrix

polystyrene/DVB

matrix active group

paired iminodiacetate groups

particle size

20-50 mesh

Application

chelating agent with high selectivity for polyvalent metal ions

Legal Information

Chelex is a registered trademark of Bio-Rad Laboratories, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service