18580
1-Bromoundecane
purum, ≥97.0% (GC)
Synonym(s):
Undecyl bromide
About This Item
grade
purum
assay
≥97.0% (GC)
refractive index
n20/D 1.457 (lit.)
n20/D 1.457
bp
137-138 °C/18 mmHg (lit.)
mp
−9 °C (lit.)
density
1.054 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCBr
InChI
1S/C11H23Br/c1-2-3-4-5-6-7-8-9-10-11-12/h2-11H2,1H3
InChI key
IKPSIIAXIDAQLG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service