Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18451

Sigma-Aldrich

3-Bromothiophenol

technical, ≥92.0% (GC)

Synonym(s):

3-Bromobenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4SH
CAS Number:
Molecular Weight:
189.07
Beilstein/REAXYS Number:
1926242
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

assay

≥92.0% (GC)

refractive index

n20/D 1.634 (lit.)
n20/D 1.634

bp

123-124 °C/40 mmHg (lit.)

density

1.832 g/mL at 25 °C (lit.)

SMILES string

Sc1cccc(Br)c1

InChI

1S/C6H5BrS/c7-5-2-1-3-6(8)4-5/h1-4,8H

InChI key

HNGQQUDFJDROPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service