Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17157

Millipore

Glucose Azide Broth

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

shelf life

limited shelf life, expiry date on the label

composition

bromocresol purple, 0.03 g/L
dextrose, 5 g/L
dipotassium phosphate, 5 g/L
monopotassium phosphate, 2 g/L
peptic digest of animal tissue, 10 g/L
sodium azide, 0.25 g/L
sodium chloride, 5 g/L
yeast extract, 3 g/L

final pH

6.7±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

Streptococcus spp.
selective for Streptococcus spp.

Application

For the enumeration of faecal Streptococci by MPN technique from water and sewage.

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service