Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16665

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside

≥98.0% (HPLC)

Synonym(s):

5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein/REAXYS Number:
1402009
EC Number:
MDL number:
UNSPSC Code:
12352204

assay

≥98.0% (HPLC)

optical activity

[α]20/D −62±2°, c = 1% in DMF/H2O 1:1

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11+,12+,13-,14-/m1/s1

InChI key

OPIFSICVWOWJMJ-AEOCFKNESA-N

Looking for similar products? Visit Product Comparison Guide

Application

X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

Other Notes

Substrate for β-galactosidase[1][2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic substrates in the histochemical demonstration of intestinal disaccharidases.
Z Lojda et al.
Histochemie. Histochemistry. Histochimie, 34(4), 361-369 (1973-03-26)
R.W. Davies et al.
Advanced Bacterial Genetics, Cold Spring Harbor (1980)
Yibing Wang et al.
PloS one, 8(3), e59195-e59195 (2013-03-26)
Our study had three objectives: to extend the plasmid-based transformation protocol to a clinical isolate of C. trachomatis belonging to the trachoma biovar, to provide "proof of principle" that it is possible to "knock out" selected plasmid genes (retaining a
Enzo R Porrello et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(1), 187-192 (2012-12-19)
We recently identified a brief time period during postnatal development when the mammalian heart retains significant regenerative potential after amputation of the ventricular apex. However, one major unresolved question is whether the neonatal mouse heart can also regenerate in response
Khanh-Van Tran et al.
Cell metabolism, 15(2), 222-229 (2012-02-14)
Adipose tissue expansion involves the enlargement of existing adipocytes, the formation of new cells from committed preadipocytes, and the coordinated development of the tissue vascular network. Here we find that murine endothelial cells (ECs) of classic white and brown fat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service