Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15392

Sigma-Aldrich

N-Boc-1,6-hexanediamine hydrochloride

purum, ≥98.0% (AT)

Synonym(s):

N-Boc-1,6-diamino-hexane hydrochloride, tert-Butyl N-(6-aminohexyl)carbamate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCONH(CH2)6NH2 · HCl
CAS Number:
Molecular Weight:
252.78
Beilstein/REAXYS Number:
3631740
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

assay

≥98.0% (AT)

reaction suitability

reagent type: cross-linking reagent

mp

162-164 °C (lit.)

SMILES string

NCCCCCCNC(OC(C)(C)C)=O.[H]Cl

InChI

1S/C11H24N2O2.ClH/c1-11(2,3)15-10(14)13-9-7-5-4-6-8-12;/h4-9,12H2,1-3H3,(H,13,14);1H

InChI key

JSBWQIZQJOQPFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the introduction of a C6-spacer.

Other Notes

Reagent for the introduction of a C6-spacer;[1][2] synthesis of polyamines and -amides[3][4]

replaced by

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.C. Yorke et al.
Australian Journal of Chemistry, 39, 447-447 (1986)
L. Kovacs et al.
Helvetica Chimica Acta, 75, 1909-1909 (1992)
J F Callahan et al.
Journal of medicinal chemistry, 32(2), 391-396 (1989-02-01)
We report the synthesis and biological activity of a series of analogues of the vasopressin antagonists [Pmp1,D-Tyr(Et)2,Val4]arginine-vasopressin (1) and [Pmp1,D-Tyr(Et)2,Val4,desGly9]arginine-vasopressin (2), where part or all of the tripeptide tail has been replaced by a simple alkyldiamine [NH(CH2)nNH2] or (aminoalkyl)guanidine [NH(CH2)nNHC(=
O. Buchardt et al.
The Journal of Organic Chemistry, 49, 4123-4123 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service