Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15247

Supelco

N,N-Bis(trimethylsilyl)formamide

technical, ≥90% (GC)

Synonym(s):

BSF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H19NOSi2
CAS Number:
Molecular Weight:
189.40
Beilstein/REAXYS Number:
2077882
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

technical

assay

≥90% (GC)

refractive index

n20/D 1.437

bp

158 °C

mp

158 °C

density

0.885 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)N(C=O)[Si](C)(C)C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the silylation of alcohols, acids and amines[1][2]; Preparation of N-TMS-imines from organolithiums[3]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N.-Y. Shih
Tetrahedron Letters, 34, 595-595 (1993)
W. Kantlehner et al.
Chemische Berichte, 105, 2264-2264 (1972)
Justus Liebigs Annalen der Chemie, 512-512 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service