10888
N-Methylacetoacetamide solution
produced by Wacker Chemie AG, Burghausen, Germany, 70% in H2O
Synonym(s):
MMAA, N-Methyl-3-oxobutyramide
About This Item
grade
produced by Wacker Chemie AG, Burghausen, Germany
concentration
70% in H2O
bp
106 °C (lit.)
density
1.101 g/mL at 20 °C (lit.)
SMILES string
CNC(=O)CC(C)=O
InChI
1S/C5H9NO2/c1-4(7)3-5(8)6-2/h3H2,1-2H3,(H,6,8)
InChI key
ATWLCPHWYPSRBQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service