Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08356

Sigma-Aldrich

(cis)-3-Aminobicyclo[2.2.1]heptane-2-carboxylic acid hydrochloride

≥98.0% (TLC)

Synonym(s):

(cis)-2-Amino-3-carboxybicyclo[2.2.1]heptane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO2 · HCl
CAS Number:
Molecular Weight:
191.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0% (TLC)

form

powder with small lumps

reaction suitability

reaction type: solution phase peptide synthesis

color

colorless to white

application(s)

peptide synthesis

SMILES string

Cl.N[C@@H]1[C@H]2CC[C@H](C2)[C@@H]1C(O)=O

InChI

1S/C8H13NO2.ClH/c9-7-5-2-1-4(3-5)6(7)8(10)11;/h4-7H,1-3,9H2,(H,10,11);1H/t4-,5+,6+,7-;/m1./s1

InChI key

GVGHCTZKPYWADE-BZUDZRPRSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service