Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07476

Supelco

1,2,3,4,7,7-Hexachloro-5-(tetrabromophenyl)bicyclo[2.2.1]hept-2-ene

analytical standard

Synonym(s):

1,2,3,4,7,7-Hexachloro-5-(tetrabromophenyl)-2-norbornene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H4Br4Cl6
CAS Number:
Molecular Weight:
692.50
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

CBr([BrH]Br)Br.ClC1=C(Cl)[C@]2(Cl)[C@@H](C3=CC=CC=C3)C[C@]1(Cl)C2(Cl)Cl

InChI

1S/C13H4Br4Cl6/c14-5-1-3(6(15)8(17)7(5)16)4-2-11(20)9(18)10(19)12(4,21)13(11,22)23/h1,4H,2H2

InChI key

PHYHWKJTYWPNCS-UHFFFAOYSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandra Catenaccio et al.
Cell death & disease, 8(11), e3166-e3166 (2017-11-10)
Wallerian degeneration is an active program tightly associated with axonal degeneration, required for axonal regeneration and functional recovery after nerve damage. Here we provide a functional molecular foundation for our undertstanding of the complex non-cell autonomous role of glial cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service