Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06453

Supelco

Amberlite LA-2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000

vapor pressure

0.01 mmHg ( 20 °C)

capacity

2.2-2.3 meq/mL capacity (sec amine)

bp

160 °C/10 mmHg (lit.)

density

0.83 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service