04390
2-Butanone peroxide
technical
Synonym(s):
Ethyl methyl ketone peroxide, Methyl ethyl ketone peroxide
About This Item
Recommended Products
grade
technical
contains
50-60% diacetone alcohol + phthalic acid ester as plasticizer
reaction suitability
reagent type: oxidant
refractive index
n20/D 1.455
density
1.053 g/mL at 20 °C (lit.)
storage temp.
2-8°C
SMILES string
CCC(C)(OO)OOC(C)(CC)OO
InChI
1S/C8H18O6/c1-5-7(3,11-9)13-14-8(4,6-2)12-10/h9-10H,5-6H2,1-4H3
InChI key
WFUGQJXVXHBTEM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Studies of the adiabatic runaway reaction of Me Et Ketone peroxide
- Quantitation of metal ions in archaeological glass via abrasive stripping square-wave voltammetry
- Imaging of hydrogen peroxide and hydrogen peroxide-scavenging substances by photon emission
- Synthesis of oligonucleotides via phosphoramidite method
- Oxidation of nucleoside phosphites into phosphates
- Comparing the relative effectiveness of human plasma glutathione peroxidase as a catalyst for reduction of hydroperoxidases
Analyte for direct electrochemical catalystis of immobilized Hb in an ethanol-water mixture
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service