Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01984

Supelco

1-Penten-3-ol

analytical standard

Synonym(s):

Ethyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(OH)CH=CH2
CAS Number:
Molecular Weight:
86.13
Beilstein/REAXYS Number:
1719833
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.424 (lit.)
n20/D 1.425

bp

114-115 °C (lit.)

density

0.838 g/mL at 20 °C (lit.)
0.839 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCC(O)C=C

InChI

1S/C5H10O/c1-3-5(6)4-2/h3,5-6H,1,4H2,2H3

InChI key

VHVMXWZXFBOANQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

82.4 °F - closed cup

flash_point_c

28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Böttcher et al.
Food chemistry, 169, 492-498 (2014-09-23)
The aim of the present study was to evaluate the off-flavour masking potential of pea dextrin (PD) in emulsions rich in ω-3 and ω-6-fatty acids in comparison with maltodextrin (MD) and 2-hydroxypropyl-β-cyclodextrin (HPBCD). After optimisation of the homogenisation procedure, stable
Tadao Wagatsuma et al.
Journal of experimental botany, 66(3), 907-918 (2014-11-25)
Several studies have shown that differences in lipid composition and in the lipid biosynthetic pathway affect the aluminium (Al) tolerance of plants, but little is known about the molecular mechanisms underlying these differences. Phospholipids create a negative charge at the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service