Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01883

Supelco

N-Succinylglycine

analytical standard

Synonym(s):

4-[(Carboxymethyl)amino]-4-oxobutanoic acid, N-(3-Carboxy-1-oxopropyl)glycine, N-(3-Carboxypropionyl)glycine, N-Glycinylsuccinamic acid, Succinyl glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO5
CAS Number:
Molecular Weight:
175.14
Beilstein/REAXYS Number:
1782053
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(CNC(CCC(O)=O)=O)=O

InChI

1S/C6H9NO5/c8-4(1-2-5(9)10)7-3-6(11)12/h1-3H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

HQESMQVYRAJTEA-UHFFFAOYSA-N

Biochem/physiol Actions

Succinylglycine is an acylglycine with a succinyl group as the acyl moiety. Acylglycines possess a common amidoacetic acid moiety and are normally minor metabolites of fatty acids. Elevated levels of certain acylglycines appear in the urine and blood of patients with various fatty acid oxidation disorders. They are normally produced through the action of glycine N-acyltransferase which is an enzyme that catalyzes the chemical reaction: acyl-CoA + glycine <-> CoA + N-acylglycine.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service