Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01510

Sigma-Aldrich

1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose

purum, ≥97.0% (TLC)

Synonym(s):

β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H24O9
CAS Number:
Molecular Weight:
504.48
Beilstein/REAXYS Number:
100243
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

assay

≥97.0% (TLC)

optical activity

[α]20/D +44±1°, c = 0.5% in chloroform

mp

128-130 °C

SMILES string

CC(=O)O[C@@H]1O[C@H](COC(=O)c2ccccc2)[C@@H](OC(=O)c3ccccc3)[C@H]1OC(=O)c4ccccc4

InChI

1S/C28H24O9/c1-18(29)34-28-24(37-27(32)21-15-9-4-10-16-21)23(36-26(31)20-13-7-3-8-14-20)22(35-28)17-33-25(30)19-11-5-2-6-12-19/h2-16,22-24,28H,17H2,1H3/t22-,23-,24-,28-/m1/s1

InChI key

GCZABPLTDYVJMP-CBUXHAPBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Ribose derivative used for the synthesis of nucleosides[1][2][3][4]; 1-cyanation with Me3SiC

replaced by

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Matsuda et al.
Synthesis, 748-748 (1981)
M. Saneyoshi et al.
Chemical & Pharmaceutical Bulletin, 27, 2518-2518 (1979)
K. Utimoto et al.
Tetrahedron Letters, 23, 237-237 (1982)
Z Tocik et al.
Nucleic acids research, 8(20), 4755-4761 (1980-10-24)
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose (I) was reacted with iodotrimethylsilane (II) and the product, the glycosyl iodide, was coupled with silylated uracil to afford 1-(2,3,5-tri-O-benzoyl-beta-D-ribofuranosyl)uracil (III; 89%), with silyated cytosine to afford, on subsequent acetylation, 1-(2,3,5-tri-O-benzoyl-beta-0D-ribofuranosyl)-4-acetamido-2-(1H)-pyrimidinone (IVb; 81%), and with chloromercuri-N-benzoyl-adenine to afford Va
A general synthesis of N-glycosides. I. Synthesis of pyrimidine nucleosides.
U Niedballa et al.
The Journal of organic chemistry, 39(25), 3654-3660 (1974-12-13)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service