Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01447

Sigma-Aldrich

Sulforhodamine 101 N-succinimidyl ester

Synonym(s):

Texas Red® N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H33N3O9S2
Molecular Weight:
703.78
MDL number:
UNSPSC Code:
12352200

CofA

specification on request

Legal Information

Texas Red is a registered trademark of Life Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service