Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00833

Sigma-Aldrich

N-Boc-hexahydro-1H-azepin-4-one

≥95.0% (HPLC)

Synonym(s):

1-Boc-homopiperazin-4-one, tert-Butyl hexahydro-4-oxo-1H-azepine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥95.0% (HPLC)

form

liquid

refractive index

n20/D 1.467

functional group

ketone

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CCCC(=O)CC1

InChI

1S/C11H19NO3/c1-11(2,3)15-10(14)12-7-4-5-9(13)6-8-12/h4-8H2,1-3H3

InChI key

PMLBUVZPRKXMOX-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service