Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

0004-PA

Sigma-Aldrich

(GN)2-3`,6`LN-C2-PAA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

carrier

PAA carrier

product line

PA-Series

assay

95% (H-NMR)

form

lyophilized powder

mol wt

~20,000 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density

technique(s)

ELISA: suitable

probe type

tetrasaccharide

probe presentation

polymeric

storage temp.

−20°C

General description

PA-Series: Unlabeled Polyacrylamide Conjugates
This series of glycoconjugate probes consists of HOCH2CH2NH3+-salts for carboxylates and sulfates. PAA is poly[N-(2-hydroxyethyl) acrylamide], a flexible polymer chain which serves as an additional spacer. The spacer-arm for oligosaccharides is normally –(CH2)3–.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Preparation Note

Purity: synthetic ω-aminoalkyl glycosides used for coupling with the polymer have a purity of >95% (HPLC and 1H-NMR)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service