Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40-0101

Sigma-Aldrich

pH standard solution

pH 1.68 (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
Pricing and availability is not currently available.

availability

available only in Japan

Quality Level

pH

1.68 (25 °C)

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service