11363727001
Roche
CAT ELISA
sufficient for 192 tests, kit of 1 (10 components), suitable for ELISA
Synonym(s):
chloramphenicol acetyltransferase
About This Item
Recommended Products
usage
sufficient for 192 tests
packaging
kit of 1 (10 components)
manufacturer/tradename
Roche
technique(s)
ELISA: suitable
storage temp.
2-8°C
General description
The bacterial enzyme chloramphenicol acetyltransferase (CAT, type I), encoded by Tn9 and having no eukaryotic equivalent, has become one of the standard markers used in transfection experiments. Traditionally, CAT activity is measured using a radioisotopic CAT assay.
Application
Features and Benefits
- Standardized: Allows direct comparison of data from different sets of experiments, even when kits from different production lots are used
- Faster: Approximately 4hours from start to finish
- Safer: No radioisotopes are used
- More accurate: Measures the actual amount of CAT protein synthesized, not just CAT enzyme activity
- Sensitive: Equivalent to the sensitivity of the radioactive CAT assay
- Easy to perform: Follows a standard ELISA protocol in microplates; allows Triton lysis of the transfected cells (CAT activity in the radioisotopic assay is rapidly inhibited by Triton)
- More flexible: Allows the analysis of promoters in plant protoplasts (alternative method to the GUS assay) and animal cells
Packaging
Principle
Preparation Note
Add 40 μl CAT enzyme stock solution (solution 1) to 3.96 ml Sample buffer (solution 7) to obtain a CAT enzyme working dilution (final concentration approx. 1 ng/ml), sufficient to produce a calibration curve in duplicate.
Handling instructions
- The CAT enzyme standard dilutions should be prepared freshly before use and should not be stored.
- Prepare the standard dilution series in reaction tubes in 1:2 dilution steps as described in the table below.
- To obtain a calibration curve, we recommend using the five concentrations listed.
- 200 μl of each dilution is needed per well.
- To ensure that the measurements and the calibration curve are accurate, we recommend preparing two samples of each concentration for measurement.
- To avoid carryover of the higher conc. solution to the lower conc. samples, use a fresh pipette tip for each dilution step.
- Each dilution must be measured in duplicate.
Storage conditions (working solution): Sample Buffer 2 to 8 °C
Stability of ready-to-use sample buffer: 2 months at 2 to 8 °C, otherwise aliquoted at - 15 to -25 °C.
Other Notes
Kit Components Only
- CAT Enzyme (type I), from E. coli
- Anti-CAT-digoxigenin antibody
- Anti-Digoxigenin-POD antibody
- POD Substrate
- Substrate Enhancer
- Washing Buffer 10x concentrated
- Sample Buffer
- Lysis Buffer 5x concentrated
- Microplates, precoated with anti-CAT (2 x strip frame with 12 modules of 8 wells)
- Self-adhesive Plate Cover Foils (3)
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Sens. 1
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 2
flash_point_f
does not flash
flash_point_c
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service