Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIEH0425E34F

Rieke Metals, Inc.

4-[(2,2,6,6-TETRAMETHYL-1-PIPERIDINO)METHYL]PHENYLZINC BROMIDE, 0.25 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
375.67
MDL number:

Iupac Name

(4-((2,2,6,6-tetramethylpiperidin-1-yl)methyl)phenyl)zinc(II) bromide

Inchi Code

1S/C16H24N.BrH.Zn/c1-15(2)11-8-12-16(3,4)17(15)13-14-9-6-5-7-10-14;;/h6-7,9-10H,8,11-13H2,1-4H3;1H;/q;;+1/p-1

InChI key

AJVCQTNQCTXYKY-UHFFFAOYSA-M

Country of Origin

US

Product Link

https://riekemetals.com/products-services/catnum/ZT2089

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library