Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIE156322646

Rieke Metals, Inc.

5-Oxo-5-(4-trifluoromethylphenyl)valeronitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
241.21
MDL number:
Product RIE156322646 is currently not available for purchase. Contact Technical Service

Iupac Name

5-oxo-5-[4-(trifluoromethyl)phenyl]pentanenitrile

Inchi Code

1S/C12H10F3NO/c13-12(14,15)10-6-4-9(5-7-10)11(17)3-1-2-8-16/h4-7H,1-3H2

InChI key

ZDPAIFYBUBUJCZ-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

Light yellow oil

Product Link

https://riekemetals.com/products-services/catnum/6046c

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library