Key Documents
OBP-5000
Oxford Lab Products
Oxford Lab BenchMate Plus Pipette
500-5000 µl, Single Channel
Select a Size
1 EA
$199.80
$199.80
Available to ship onMay 08, 2025Details
Select a Size
Change View
1 EA
$199.80
About This Item
PDF Icon
Shipped & Sold By Oxford Lab Products
A Trusted Marketplace Seller
PDF Icon
Have a question?
$199.80
Available to ship onMay 08, 2025Details
manufacturer/tradename
Oxford Lab Products
General description
Oxford Lab BenchMate Plus Pipette
The Oxford BenchMate Plus Pipette offers an incredibly smooth and lightweight experience that optimizes precision for the across all users and for full days of use!
Features
  • Modern, lightweight materials enable a durable design without burdening the user. The Oxford BenchMate Plus pipette offers a very lightweight body.
  • The Oxford BenchMate Plus is designed with the classic BenchMate pipette body and additionally equipped with a smooth and round fingerhook. The classic, yet modern, design delivers an incredibly ergonomic experience.
  • An internal magnetic system and smooth plunger design yield an incredibly soft and smooth plunger force that allows users to easily find the first and second stop. This system yields a higher precision based on the user technique rather than just mechanical specifications.
  • Oxford BenchMate Plus Pipettes are color-coded according to their size range, making it easy to identify the pipette size, even from a distance.
  • The new Oxford BenchMate Plus Pipette volume lock design features an easy tab that only requires a flip from side-to-side to lock or unlock the volume adjustment.
  • Oxford Benchmate Plus Multichannel Pipettes feature an O-ring installed onto the tip cones, making it easier for users to effectively attach pipette tips without needing excessive force. The O-Rings also enable more secure attachment vs regular tip cones.
Properties
  • Volume Lock secure volume lock prevents accidental volume lock
  • Magnetic Piston System Internal magnets guide user for added precision
  • Lightweight Body light materials and design optimize ergonomic design
  • Spring-loaded tip cone reduces force needed to achieve effective tip attachment
  • Very Light and Smooth Plunger minimal plunging force optimizes user precision
  • Fully Autoclavable entire pipette can be autoclaved for easy cleaning
  • O-Ring Equiped Multichannel Tips o-ring designs enables easier and more secure tip attachment
The Oxford BenchMate Plus Pipette offers all the tangibles and intangibles needed to be a premium pipette. Mechanically, the BenchMate Plus is made with durable and comfortable plastics that offer an easy grip for the user, a lightweight experience for ergonomics, and a resistance to chemical or physical damage. Inside, all pieces are finely honed and designed to be incredibly smooth and fit perfectly, creating a premium fit and long-lasting product life. This incredible care and design is obvious when using the BenchMate Plus, particularly in the smooth plunger with minimal piston forces.
Above all mechanical specifications, the Oxford BenchMate Plus offers a highly ergonomic experience that enables uses to be accurate and precise beyond the simple hardware limitations. The internal magnet system and minimal piston force enables users to maintain consistent and repetitive plunging motions, thus affording advanced precision beyond the mechanical specifications. Furthermore, the Oxford BenchMate Plus ergonomic design means that users can feel comfortable and relaxed for longer, avoiding stress injuries and variability.
Choose from one of the most recent versions:
Certificates of Analysis (COA)