Key Documents
OB-200
Oxford Lab Products
Oxford Lab BenchMate Pipette
20-200 µl
About This Item
PDF Icon
Shipped & Sold By Oxford Lab Products
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Oxford Lab Products
General description
BenchMate Model Autoclavable Single Channel Pipette
Durable, quality alternative to Gilson Classic pipette design. Easy and convenient single hand locking lever. Eight sizes to cover a wide volume range from 0.1µL to 10mL. PTFE Plastic used in combination with proprietary technology to ensure consistent air travel for precise and accurate pipetting.
Features
  • Easy to use single handed operation with a locking mechanism
  • Fully Autoclavable (121°C for 20 minutes)
  • Improved dispensing accuracy and precision for micro volumes between 2µL and 10µL with hyper blow out technology
  • Ceramic plungers for greater durability for pipettes ranging from 200µL to 10mL
  • Eight sizes to cover a wide volume range from 0.1µL to 10mL
  • PTFE Plastic used in combination with proprietary technology to ensure consistent air travel for precise and accurate pipetting
  • Enhanced UV resistance for under the hood work
  • 5 Year Manufacturer Warranty