Key Documents
19-353-0150
Omni International
Omni Bead Ruptor 12 1.5mL Tube Adapter kit
About This Item
PDF Icon
Shipped & Sold By Omni International
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Omni International
General description
Adapter Kit for 1.5 mL Microcentrifuge Tube. Each adapter snaps into a 7 mL tube processing position on the Bead Ruptor 12 carriage providing a 4 x 1.5 mL processing capability.
Choose from one of the most recent versions:
Certificates of Analysis (COA)