OM-130810
OMICRON™
OMICRON™ Glass Fiber Filters
Borosilicate Glass Fiber Binder Free Filter, 1.6 Micron, 8"X10" (203mm X 254mm), (100/Pk)
Synonym(s):
Glass Fiber, Filters, GF/A, Membrane
Select a Size
$181.00
In StockDetails
Select a Size
About This Item
$181.00
In StockDetails
Recommended Products
manufacturer/tradename
Omicron Scientific
General description
OMICRON™ Glass Fiber filters are manufctured from pure Borosilicate Glass Fibers and are available in a wide range of nominal pore sizes. These filters have extremely low pressure drop, making them an excellent choice as a pre-filter for fluids that have high viscous and high dust holding capacity. Target applications include salt, sugar, sera, tissue lysates, protein solutions, and environmental samples, like soil and ground water, where there is an elevated level of suspended or un-dissloved particles. The production process of OMICRON™ Glass Fiber Filters has been validated with an accredited registered body to ISO 9001:2015 Quality System Standards.
Features and Benefits- Biologically inert
- High dirt loading capacity
- Available with Acrylic binders
- Low resistance with high filtration efficiency
- Hydrophilic material membrane
- Low Extractable
- Stack sampling and Air pollution monitoring
- Water pollution monitoring of effluents
- Filtration of water, algae and bacteria cultures
- Foodstuff analysis
- Gravimetric determination of airborne particulates
(Cross Reference GF/A 1820-866)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)